Learn More
As One International Inc ELEP CLEANER A-3500 160
Catalog No. NC2606701
Encompass
  • Excellent dust removal performance
  • Firmly captures foreign materials
  • Made of PET non-woven fabric
  • Acrylic adhesive type
  • Core diameter of 38 mm
  • 160 mm width
  • Length of 20 m
  • Easy-to-cut perforated design
  • Ideal for cleaning floors, workbenches, and apparel
  • RoHS3 (EU) 2015/863 compliant
  • Suitable for high cleanliness environments like laboratories and cleanrooms
Catalog No. NC2606701 Supplier As One International Inc Supplier No. 9501002
  • Excellent dust removal performance
  • Firmly captures foreign materials
  • Made of PET non-woven fabric
  • Acrylic adhesive type
  • Core diameter of 38 mm
  • 160 mm width
  • Length of 20 m
  • Easy-to-cut perforated design
  • Ideal for cleaning floors, workbenches, and apparel
  • RoHS3 (EU) 2015/863 compliant
  • Suitable for high cleanliness environments like laboratories and cleanrooms