Learn More
Selleck Chemical LLC Elesclomol (STA-4783) S1052-200mg
SDP
Catalog No. 503108212
Encompass_Preferred
Elesclomol (STA-4783) is a novel potent oxidative stress inducer that elicits pro-apoptosis events among tumor cells Phase 3 Elesclomol specifically binds ferredoxin 1 (FDX1) 2/ 3 helices and 5 strand and inhibits FDX1-mediated Fe-S cluster biosynthesis Elesclomol (STA-4783) is a potent copper ionophore and can be used in the research of copper-dependent cell death (cuproptosis)
Catalog No. 50-310-8212 Supplier Selleck Chemical LLC Supplier No. S1052200MG
Elesclomol (STA-4783) is a novel potent oxidative stress inducer that elicits pro-apoptosis events among tumor cells Phase 3 Elesclomol specifically binds ferredoxin 1 (FDX1) 2/ 3 helices and 5 strand and inhibits FDX1-mediated Fe-S cluster biosynthesis Elesclomol (STA-4783) is a potent copper ionophore and can be used in the research of copper-dependent cell death (cuproptosis)