Learn More
TARGETMOL CHEMICALS INC ELESCLOMOL 50MG
SDP
Catalog No. 502576583
Encompass_Preferred
Also available in 5 mg 10 mg 25 mg 100 mg 200 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Elesclomol (STA-4783) (STA-4783) is an effective oxidative stress inducer that elicits pro-apoptosis of tumor cells. purity: 99%
Catalog No. 50-257-6583 Supplier TARGETMOL CHEMICALS INC Supplier No. T617050MG
Also available in 5 mg 10 mg 25 mg 100 mg 200 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Elesclomol (STA-4783) (STA-4783) is an effective oxidative stress inducer that elicits pro-apoptosis of tumor cells. purity: 99%