Learn More
Apexbio Technology LLC Elesclomol (STA-4783),5mg CAS# 488832-69-5
SDP
Catalog No. 502240099
Encompass_Preferred
Elesclomol (also known as STA-4783), originally identified in a cell-based phenotypic screen for proapoptotic activity, is a novel small-molecule that potently induces apoptosis of cancer cells through the rapid generation of reactive oxygen species (ROS) and the induction of unmanageable levels of oxidative stress. Elesclomol exhibits autitumor activity against a broad spectrum of types of cancer cell in human tumor xengograft models due to its excessive ROS production and elevated levels of oxidative stress leading to the death of cancer cells.  Other sizes are also available. Please inqury us for quote.
This item is not returnable. View return policy
Catalog No. 50-224-0099 Supplier Apexbio Technology LLC Supplier No. A43865
Elesclomol (also known as STA-4783), originally identified in a cell-based phenotypic screen for proapoptotic activity, is a novel small-molecule that potently induces apoptosis of cancer cells through the rapid generation of reactive oxygen species (ROS) and the induction of unmanageable levels of oxidative stress. Elesclomol exhibits autitumor activity against a broad spectrum of types of cancer cell in human tumor xengograft models due to its excessive ROS production and elevated levels of oxidative stress leading to the death of cancer cells.  Other sizes are also available. Please inqury us for quote.