Learn More
Medchemexpress LLC Eletriptan hydrobromide | 177834-92-3 | 99.8% | C22H27BrN2O2S | 100 MG
SDP
Catalog No. 5000433215
Encompass_Preferred
Eletriptan hydrobromide is a 5-HT1B and 5-HT1D receptor agonist with antimigraine activity, exhibiting Ki values of 0.92 nM and 3.14 nM, respectively. It is intended for research use only. In vivo studies have shown that eletriptan hydrobromide dose-dependently reduces carotid blood flow in anesthetized dogs and inhibits dural plasma protein extravasation in anesthetized rats.
  • Functions as a 5-HT1B and 5-HT1D receptor agonist.
  • Exhibits Ki values of 0.92 nM for 5-HT1B receptor.
  • Exhibits Ki values of 3.14 nM for 5-HT1D receptor.
  • Dose-dependently reduces carotid blood flow in anesthetized dogs.
  • Inhibits dural plasma protein extravasation in anesthetized rats.
  • Completely inhibits plasma protein extravasation from the dura mater at 100 μg/kg.
Catalog No. 50-004-33215 Supplier Medchemexpress LLC Supplier No. HYA0010100MG
Eletriptan hydrobromide is a 5-HT1B and 5-HT1D receptor agonist with antimigraine activity, exhibiting Ki values of 0.92 nM and 3.14 nM, respectively. It is intended for research use only. In vivo studies have shown that eletriptan hydrobromide dose-dependently reduces carotid blood flow in anesthetized dogs and inhibits dural plasma protein extravasation in anesthetized rats.
  • Functions as a 5-HT1B and 5-HT1D receptor agonist.
  • Exhibits Ki values of 0.92 nM for 5-HT1B receptor.
  • Exhibits Ki values of 3.14 nM for 5-HT1D receptor.
  • Dose-dependently reduces carotid blood flow in anesthetized dogs.
  • Inhibits dural plasma protein extravasation in anesthetized rats.
  • Completely inhibits plasma protein extravasation from the dura mater at 100 μg/kg.