Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Eletriptan hydrobromide | 177834-92-3 | MFCD08141806 | 463.43 g/mol | C22H27BrN2O2S | 10 MG
SDP

Catalog No. 5000298442
Encompass_Preferred

Eletriptan hydrobromide is the hydrobromide salt of eletriptan, a selective 5-HT1B and 5-HT1D receptor agonist supplied as an analytical standard for laboratory and research use.

  • Analytical standard suitable for research and assay development.
  • Selective 5-HT1B and 5-HT1D agonist for pharmacology studies.
  • Reported Ki values: 0.92 nM (5-HT1B) and 3.14 nM (5-HT1D).
  • Chemical formula C22H27BrN2O2S; molecular weight ~463.4 g/mol.
  • Provided in small masses appropriate for analytical work.

Catalog No. 50-002-98442 Supplier Medchemexpress LLC Supplier No. HYA001010MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Eletriptan hydrobromide is the hydrobromide salt of eletriptan, a selective 5-HT1B and 5-HT1D receptor agonist supplied as an analytical standard for laboratory and research use.

  • Analytical standard suitable for research and assay development.
  • Selective 5-HT1B and 5-HT1D agonist for pharmacology studies.
  • Reported Ki values: 0.92 nM (5-HT1B) and 3.14 nM (5-HT1D).
  • Chemical formula C22H27BrN2O2S; molecular weight ~463.4 g/mol.
  • Provided in small masses appropriate for analytical work.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.