Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Eleutheroside B1 | 16845-16-2 | MFCD32173675 | 98.7% | 384.33 g/mol | C17H20O10 | 5 MG
SDP

Catalog No. 5000222852
Encompass_Preferred

Eleutheroside B1 is a naturally occurring coumarin glycoside provided as a purified reference compound for biochemical and pharmacological research. It has reported in vitro anti-influenza activity and is supplied as a small-quantity analytical standard for laboratory studies.

  • High purity (98.7% reported)
  • Molecular formula C17H20O10
  • Molecular weight 384.33 g/mol
  • Registered CAS number 16845-16-2
  • Supplied as a small vial for research use only
  • White to off-white solid, suitable for analytical assays

Catalog No. 50-002-22852 Supplier Medchemexpress LLC Supplier No. HY1356465MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Eleutheroside B1 is a naturally occurring coumarin glycoside provided as a purified reference compound for biochemical and pharmacological research. It has reported in vitro anti-influenza activity and is supplied as a small-quantity analytical standard for laboratory studies.

  • High purity (98.7% reported)
  • Molecular formula C17H20O10
  • Molecular weight 384.33 g/mol
  • Registered CAS number 16845-16-2
  • Supplied as a small vial for research use only
  • White to off-white solid, suitable for analytical assays

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.