Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Eleutheroside C | 15486-24-5 | 1 MG
SDP

Catalog No. 5000419266
Encompass_Preferred

Eleutheroside C (Ethyl α-D-galactoside) is a glycoside isolated from the bulbs of *Polianthes tuberosa*. It has a molecular weight of 208.21 and a chemical formula of C8H16O6. This white to off-white solid is classified as a saccharide and originates from plants in the Araliaceae family, specifically *Acanthopanax senticosus (Rupr. et Maxim.) Harms*.

  • Purity: 99.90%
  • For research use only
  • Shipped at room temperature in continental US
  • Store at 4°C, protected from light
  • Store in solvent at -80°C for 6 months or -20°C for 1 month, protected from light

Catalog No. 50-004-19266 Supplier Medchemexpress LLC Supplier No. HYN38021MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Eleutheroside C (Ethyl α-D-galactoside) is a glycoside isolated from the bulbs of *Polianthes tuberosa*. It has a molecular weight of 208.21 and a chemical formula of C8H16O6. This white to off-white solid is classified as a saccharide and originates from plants in the Araliaceae family, specifically *Acanthopanax senticosus (Rupr. et Maxim.) Harms*.

  • Purity: 99.90%
  • For research use only
  • Shipped at room temperature in continental US
  • Store at 4°C, protected from light
  • Store in solvent at -80°C for 6 months or -20°C for 1 month, protected from light

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.