Learn More
Apexbio Technology LLC ELEUTHEROSIDE-E-20MG
SDP
Catalog No. 5000567756
Encompass_Preferred
Eleutheroside E (CAS No 39432-56-9) is a bioactive compound primarily studied in the field of phytochemistry and pharmacology due to its potential health benefits It originates from the Eleutherococcus senticosus plant commonly known as Siberian ginseng The compound is known to interact with multiple biological pathways including antioxidant and anti-inflammatory pathways that may influence cellular stress and immune responses Eleutheroside E exhibits activity typically in the nanomolar to low micromolar range in in vitro studies making it a candidate for further drug development and screening Its applications include various cell models to study neuroprotective and cardioprotective effects as well as animal studies investigating its potential as an adaptogen Experimental concentrations typically depend on the specific research design and its effects continue to be a subject of ongoing biomedical research
Catalog No. 50-005-67756 Supplier Apexbio Technology LLC Supplier No. N163320
Eleutheroside E (CAS No 39432-56-9) is a bioactive compound primarily studied in the field of phytochemistry and pharmacology due to its potential health benefits It originates from the Eleutherococcus senticosus plant commonly known as Siberian ginseng The compound is known to interact with multiple biological pathways including antioxidant and anti-inflammatory pathways that may influence cellular stress and immune responses Eleutheroside E exhibits activity typically in the nanomolar to low micromolar range in in vitro studies making it a candidate for further drug development and screening Its applications include various cell models to study neuroprotective and cardioprotective effects as well as animal studies investigating its potential as an adaptogen Experimental concentrations typically depend on the specific research design and its effects continue to be a subject of ongoing biomedical research