Learn More
Stoelting™ Elevated Plus Maze
SDP
Catalog No. 10000302 Shop All Stoelting Co Products
Change view
Click to view available options
Animal:
Mouse
Rat
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Animal
10-000-302 Mouse
10-000-323 Rat
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 10-000-302 Supplier Stoelting™ Supplier No. 60140
Add to Cart
Add to Cart
Stoelting™ Elevated Plus-Maze is used to assess anxiety-like behavior in laboratory animals.
  • Maze exploits the conflict between the innate fear that rodents have of open areas versus their desire to explore novel environments
  • Open Arms are equipped with a height-adjustable lip to minimize falls
  • Lip or ledge can be adjusted from 0 to 15 mm to minimize falls and to provide optional thigmotactic cues
  • Available for rat or mouse
  • Closed arm walls are held solidly in slotted base, but lift off for easy cleaning
  • Grey, non reflective base plate
  • Removable walls and legs mean the maze can be stored flat when not in used
Specifications
For Use With (Application) Neuroscience
Animal Mouse