Learn More
IONBENCH CORPORATION Elevating Ionbench LC for HPLC Worksurface size 26.5" x 25.5" (67 x 65 cm), two columns, max weight 310 lbs (140 kg), display with 3 positions memorization
Catalog No. 5000150571
Encompass_Preferred
Elevating Ionbench LC for HPLC Worksurface size 26.5" x 25.5" (67 x 65 cm), two columns, max weight 310 lbs (140 kg), display with 3 positions memorization. Optional features :DR_LC1 One drawer under the worksurface,KEY_LOCK Automatic locking digital height controller Prevent involuntary bench height movement. IEC IEC power strip including extention IEC cords Fixed underneath bench worksurface. SOLV Container for solvent waste bottles.CAN-5L Solvent waste canister 1.3 US gal. (5 liters) with safety cap Compatible with p/n SOLV & LAT-SOLV container. SOLV-MNFLD Magnetic manifold to collect solvent waste tubing With connections and hose between manifold and canister. EXH-FIL Exhaut filter for CAN-10L canister Charcoal filter (100 grams), service life 6 month. ELEC-LEVEL Solvent waste level detector with electronic module Alarm sound when canister is full Disc sensor with Velcro fastener to be placed on the canister (no contact technology)
Catalog No. 50-001-50571 Supplier IONBENCH CORPORATION Supplier No. LC2625INCHCARE
Elevating Ionbench LC for HPLC Worksurface size 26.5" x 25.5" (67 x 65 cm), two columns, max weight 310 lbs (140 kg), display with 3 positions memorization. Optional features :DR_LC1 One drawer under the worksurface,KEY_LOCK Automatic locking digital height controller Prevent involuntary bench height movement. IEC IEC power strip including extention IEC cords Fixed underneath bench worksurface. SOLV Container for solvent waste bottles.CAN-5L Solvent waste canister 1.3 US gal. (5 liters) with safety cap Compatible with p/n SOLV & LAT-SOLV container. SOLV-MNFLD Magnetic manifold to collect solvent waste tubing With connections and hose between manifold and canister. EXH-FIL Exhaut filter for CAN-10L canister Charcoal filter (100 grams), service life 6 month. ELEC-LEVEL Solvent waste level detector with electronic module Alarm sound when canister is full Disc sensor with Velcro fastener to be placed on the canister (no contact technology)