Learn More
Roboz Surgical ELEVATOR
SDP
Catalog No. NC9409697
Encompass
Freer Elevator; Double Ended; Sharp blade:1.0mm thick; Blunt blade: 1.5mm thick; 5mm Width; 7 in. Length
Catalog No. NC9409697 Supplier Roboz Surgical Supplier No. RS8820
Freer Elevator; Double Ended; Sharp blade:1.0mm thick; Blunt blade: 1.5mm thick; 5mm Width; 7 in. Length