Learn More
Med Vet International Miltex Dental 46 Elevator, Stainless Steel
Catalog No. 502875560
Encompass_Preferred
The Miltex DEL46 46 elevator is made of OR grade German stainless steel. It features: Stainless steel tip Permanently welded steel handle- no seam to crack and no plating to peel or discolor Convenient finger platforms for proper alignment of hands on the handles All-stainless steel handle- twice the strength of ordinary brass and chrome-plated handles Elevators are used to loosen and elevate teeth in their sockets prior to extraction in order to create space and prevent trauma to adjacent teeth and tissues.
Catalog No. 50-287-5560 Supplier Med Vet International Supplier No. MILDEL46
The Miltex DEL46 46 elevator is made of OR grade German stainless steel. It features: Stainless steel tip Permanently welded steel handle- no seam to crack and no plating to peel or discolor Convenient finger platforms for proper alignment of hands on the handles All-stainless steel handle- twice the strength of ordinary brass and chrome-plated handles Elevators are used to loosen and elevate teeth in their sockets prior to extraction in order to create space and prevent trauma to adjacent teeth and tissues.