Learn More
Med Vet International Miltex Molt Periosteal Elevator, Size 9, Double-Ended, 5mm And 9mm Wide, Delm9
Catalog No. 502875565
Encompass_Preferred
The surgical “feel” of these instruments, the outstanding cutting ability and the attention to delicate detail are just a few of the aspects that allow Miltex to outperform and stand out from the competition. This is a Miltex Molt Periosteal Elevator. Elevators are instruments used in dental extractions or surgical procedures. In dental procedures they may be used to loosen teeth prior to forcep extraction, to remove roots or impacted teeth, when teeth are compromised and susceptible to fracture or when they are malpositioned and cannot be reached with forceps. In surgical procedures they can be used for scraping, elevating or dissecting bones or tissues. They are made from premium-grade surgical stainless steel. Specifications: Size 9, Double-Ended, 5mm And 9mm Wide, DELM9
Catalog No. 50-287-5565 Supplier Med Vet International Supplier No. MILDELM9
The surgical “feel” of these instruments, the outstanding cutting ability and the attention to delicate detail are just a few of the aspects that allow Miltex to outperform and stand out from the competition. This is a Miltex Molt Periosteal Elevator. Elevators are instruments used in dental extractions or surgical procedures. In dental procedures they may be used to loosen teeth prior to forcep extraction, to remove roots or impacted teeth, when teeth are compromised and susceptible to fracture or when they are malpositioned and cannot be reached with forceps. In surgical procedures they can be used for scraping, elevating or dissecting bones or tissues. They are made from premium-grade surgical stainless steel. Specifications: Size 9, Double-Ended, 5mm And 9mm Wide, DELM9