Learn More
Medchemexpress LLC Elevenostat | 1454902-97-6 | 95.1% | C16H17N3O4 | 100 MG
SDP
Catalog No. 5000354384
Encompass_Preferred
Elevenostat is a selective HDAC11 inhibitor with an IC50 of 0.235 μM. It can induce apoptosis of multiple myeloma cells and has an anti-tumor effect. Additionally, it inhibits the maturation of mouse oocytes.
  • Selective HDAC11 inhibitor with an IC50 of 0.235 μM
  • Induces apoptosis in multiple myeloma cells
  • Exhibits anti-tumor effects
  • Inhibits maturation of mouse oocytes
Catalog No. 50-003-54384 Supplier Medchemexpress LLC Supplier No. HY145757100MG
Elevenostat is a selective HDAC11 inhibitor with an IC50 of 0.235 μM. It can induce apoptosis of multiple myeloma cells and has an anti-tumor effect. Additionally, it inhibits the maturation of mouse oocytes.
  • Selective HDAC11 inhibitor with an IC50 of 0.235 μM
  • Induces apoptosis in multiple myeloma cells
  • Exhibits anti-tumor effects
  • Inhibits maturation of mouse oocytes