Learn More
Medchemexpress LLC Elexacaftor-d3 (VX-445-d3) | 2216712-66-0 | 98.3% | 600.67 | 1 MG
SDP
Catalog No. 5000439230
Encompass_Preferred
Elexacaftor-d3 (VX-445-d3) is a deuterium labeled compound, serving as a tracer and internal standard for quantitative analysis methods such as NMR, GC-MS, or LC-MS. Deuteration may influence the pharmacokinetic and metabolic profiles of drugs. Its unlabeled counterpart, Elexacaftor (VX-445), acts as a next-generation cystic fibrosis transmembrane conductance regulator (CFTR) corrector, aiming to restore Phe508del CFTR protein function and treat cystic fibrosis.
  • Deuterium labeled compound
  • Used as a tracer
  • Used as an internal standard for quantitative analysis
  • Potential to affect pharmacokinetic and metabolic profiles of drugs
  • Restores Phe508del CFTR protein function
  • Has potential to treat cystic fibrosis
  • Improves Phe508del CFTR protein processing, trafficking, and chloride transport
Catalog No. 50-004-39230 Supplier Medchemexpress LLC Supplier No. HY111772S11MG
Elexacaftor-d3 (VX-445-d3) is a deuterium labeled compound, serving as a tracer and internal standard for quantitative analysis methods such as NMR, GC-MS, or LC-MS. Deuteration may influence the pharmacokinetic and metabolic profiles of drugs. Its unlabeled counterpart, Elexacaftor (VX-445), acts as a next-generation cystic fibrosis transmembrane conductance regulator (CFTR) corrector, aiming to restore Phe508del CFTR protein function and treat cystic fibrosis.
  • Deuterium labeled compound
  • Used as a tracer
  • Used as an internal standard for quantitative analysis
  • Potential to affect pharmacokinetic and metabolic profiles of drugs
  • Restores Phe508del CFTR protein function
  • Has potential to treat cystic fibrosis
  • Improves Phe508del CFTR protein processing, trafficking, and chloride transport