Learn More
Aviva Systems Biology Corp ELF1 Antibody - N-terminal region (ARP36845_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36845_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ELF1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of mouse ELF1. Alternate names: p70, Sts1, Elf-1, mElf-1ApplicationWB. Species reactivity: Dog, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-56283