Learn More
Aviva Systems Biology Corp ELF3 Antibody - middle region (ARP32247_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32247_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against ELF3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ELF3. Alternate names: ERT, ESX, EPR-1, ESE-1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-54496