Learn More
Aviva Systems Biology Corp Elf3 Antibody - N-terminal region (ARP36846_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36846_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Elf3. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of mouse Elf3. Alternate names: ESE-1, ESX, jenApplicationIHC, WB. Species reactivity: Mouse, Rat
Catalog No. 50-014-56284