Learn More
Aviva Systems Biology Corp Elf3 Antibody - middle region (ARP31639_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP31639_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ELF3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human Elf3. Alternate names: EPR-1, ERT, ESE-1, ESXApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Pig, Rabbit
Catalog No. 50-014-54291