Learn More
Aviva Systems Biology Corp Elf3 Antibody - middle region (ARP40156_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40156_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Elf3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of mouse Elf3. Alternate names: ESE-1, ESX, jenApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-57307