Learn More
Aviva Systems Biology Corp ELF4 Antibody - C-terminal region (ARP32248_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32248_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ELF4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human ELF4. Alternate names: ELFR, MEFApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-54497