Learn More
Aviva Systems Biology Corp ELF4 Antibody - N-terminal region (ARP31640_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP31640_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ELF4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ELF4. Alternate names: ELFR, MEFApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-54292