Learn More
Aviva Systems Biology Corp Elf5 Antibody - middle region (ARP37035_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37035_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Elf5. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of mouse Elf5. Alternate names: ESE-2, ESE-5., ese-5, ESE-5ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-284-569