Learn More
Aviva Systems Biology Corp ELFN2 Antibody - N-terminal region (ARP50215_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP50215_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ELFN2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ELFN2. Alternate names: KIAA1904, LRRC62, dJ63G5.3, PPP1R29ApplicationWB. Species reactivity: Cow, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-60287