Learn More
World Precision Instrument Elgiloy-Stainless Electrode Profile C-SSM123B20KT
SDP
Catalog No. 502538561
Encompass
This Elgiloy-Stainless steel electrode is used for specialty applications. The electrode provides good reading performance in acute and chronic applications, and allows for visual identification of the electrode tracts when using Prussian blue staining techniques.
Catalog No. 50-253-8561 Supplier World Precision Instrument Supplier No. SSM123B20KT
This Elgiloy-Stainless steel electrode is used for specialty applications. The electrode provides good reading performance in acute and chronic applications, and allows for visual identification of the electrode tracts when using Prussian blue staining techniques.