Learn More
World Precision Instrument Elgiloy-Stainless Electrode Profile A-SSM33A20KT
SDP
Catalog No. 502538554
Encompass
Elgiloy is a stainless steel alloy with a cobalt-chromium component. It is used for monopolar electrodes for specialty applications, and is frequently found in dental implants. Its biocompatibility is good, and it is corrosion resistant.
Catalog No. 50-253-8554 Supplier World Precision Instrument Supplier No. SSM33A20KT
Elgiloy is a stainless steel alloy with a cobalt-chromium component. It is used for monopolar electrodes for specialty applications, and is frequently found in dental implants. Its biocompatibility is good, and it is corrosion resistant.