Learn More
Medchemexpress LLC Eliapixant | 1948229-21-7 | 99.95% | 478.49 | 100 MG
SDP
Catalog No. 5000344126
Encompass_Preferred
Eliapixant (BAY 1817080) is a potent and selective antagonist of the P2X3 receptor, exhibiting an IC50 of 8 nM. This compound is utilized in research concerning refractory chronic cough. Additionally, Eliapixant can be employed for the study of coughs and complications associated with diabetes.
  • Potent and selective antagonist of P2X3 receptor, with an IC50 of 8 nM.
  • Used for the research of refractory chronic cough.
  • Applicable for research into coughs and diabetes complications.
Catalog No. 50-003-44126 Supplier Medchemexpress LLC Supplier No. HY10917050MG
Eliapixant (BAY 1817080) is a potent and selective antagonist of the P2X3 receptor, exhibiting an IC50 of 8 nM. This compound is utilized in research concerning refractory chronic cough. Additionally, Eliapixant can be employed for the study of coughs and complications associated with diabetes.
  • Potent and selective antagonist of P2X3 receptor, with an IC50 of 8 nM.
  • Used for the research of refractory chronic cough.
  • Applicable for research into coughs and diabetes complications.