Learn More
Medchemexpress LLC Eliapixant (BAY 1817080) | 1948229-21-7 | 99.9% | 478.49 | 1 ML
SDP
Catalog No. 5000345385
Encompass_Preferred
Eliapixant is a potent and selective antagonist of the P2X3 receptor, with an IC50 of 8 nM. It is used for research on refractory chronic cough and can also be applied to research on coughs and diabetes complications. This compound has been involved in clinical trials for various conditions.
  • Potent and selective antagonist of P2X3 receptor with an IC50 of 8 nM.
  • Used for research on refractory chronic cough.
  • Applicable for research on coughs and diabetes complications.
  • Undergoing clinical trials for conditions including endometriosis related pain, overactive bladder, diabetic neuropathic pain, and refractory or unexplained chronic cough.
Catalog No. 50-003-45385 Supplier Medchemexpress LLC Supplier No. HY10917010MG
Eliapixant is a potent and selective antagonist of the P2X3 receptor, with an IC50 of 8 nM. It is used for research on refractory chronic cough and can also be applied to research on coughs and diabetes complications. This compound has been involved in clinical trials for various conditions.
  • Potent and selective antagonist of P2X3 receptor with an IC50 of 8 nM.
  • Used for research on refractory chronic cough.
  • Applicable for research on coughs and diabetes complications.
  • Undergoing clinical trials for conditions including endometriosis related pain, overactive bladder, diabetic neuropathic pain, and refractory or unexplained chronic cough.