Learn More
Medchemexpress LLC HY-14885 100mg , Eliglustat Genz 99067 CAS:491833-29-5 Purity:98%
SDP
Catalog No. 501493090
Encompass_Preferred
Medchemexpress, HY-14885 100mg Eliglustat Genz 99067 CAS:491833-29-5 Formula:C23H36N2O4 IC50: 24 nM (glucocerebroside synthase) Purity:98% Eliglustat is an specific, potent and orally active glucocerebroside synthase inhibitor with an IC 50 of 24 nM. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-3090 Supplier Medchemexpress LLC Supplier No. HY14885100MG
Medchemexpress, HY-14885 100mg Eliglustat Genz 99067 CAS:491833-29-5 Formula:C23H36N2O4 IC50: 24 nM (glucocerebroside synthase) Purity:98% Eliglustat is an specific, potent and orally active glucocerebroside synthase inhibitor with an IC 50 of 24 nM. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.