Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nilfisk™ Eliminator Pro II Light Industrial Vacuum

Catalog No. 19022439 Shop All Nilfisk of America Products
Change view
Click to view available options
Filter Type:
HEPA
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Filter Type
19-022-439 HEPA
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-022-439 Supplier Nilfisk™ Supplier No. 107412640
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The Eliminator Pro II offers wet and dry collection in a light and maneuverable single-phase vacuum.

  • A true wet and dry vacuum, the Eliminator can collect dust or up to 12 gallons of water
  • Each Eliminator Pro II vacuum comes standard with a cloth main filter, motor filter, and an upstream HEPA
  • Easy to service, and even easier to use
  • Optional 24 in. wide fixed floor nozzle allows the operator to pick up spills quickly and efficiently
  • Swivel casters, large rear wheels and handle make maneuvering around obstacles or over uneven surfaces painless
  • When full, the motorhead is simply removed and the 7 gallon cloth dust bag disposed of
  • If used to collect water, there is a handy drain hose at the rear of the tank that makes emptying very easy
  • Standard HEPA filtration 50 ft. power cord
  • Upstream HEPA Available 24 in. wide fixed floor nozzle
  • Quiet operation
  • Easy to use and service 8 ft. hose, stainless steel 2-piece extension wand and accessories included

Specifications

Voltage 110/120 V
Product Type Industrial Vacuum
For Use With (Equipment) HEPA Filter System
Filter Type HEPA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.