Learn More
Medchemexpress LLC HY-101566 5mg Medchemexpress, Elimusertib CAS:1876467-74-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1015665MG
Encompass_Preferred
Medchemexpress, HY-101566 5mg Elimusertib CAS:1876467-74-1 Elimusertib (BAY-1895344) is a potent, orally available and selective ATR inhibitor with an IC50 of 7 nM. Anti-tumor activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0639