Learn More
Medchemexpress LLC Elimusertib (BAY 1895344) | 1876467-74-1 | 99.7% | 375.43 | 100 MG
SDP
Catalog No. 5000364916
Encompass_Preferred
Elimusertib (BAY-1895344) is a potent, orally active, and selective ATR inhibitor with an IC50 of 7 nM. It demonstrates anti-tumor activity and is suitable for research into solid tumors and lymphomas.
  • Potently inhibits proliferation of human tumor cell lines.
  • Suppresses hydroxyurea-induced H2AX phosphorylation.
  • Exhibits high selectivity against mTOR, DNA-PK, ATM, and PI3K.
  • Shows potent antiproliferative activity against various cancer cell lines.
  • Demonstrates potent anti-tumor efficacy in monotherapy in xenograft models of ovarian and colorectal cancer.
  • Causes complete tumor remission in mantle cell lymphoma models.
  • Has synergistic antitumor activity when combined with Carboplatin in platinum-resistant human CRC PDX models.
  • Exhibits moderate oral bioavailability and short elimination half-lives.
Catalog No. 50-003-64916 Supplier Medchemexpress LLC Supplier No. 100MGHY101566
Elimusertib (BAY-1895344) is a potent, orally active, and selective ATR inhibitor with an IC50 of 7 nM. It demonstrates anti-tumor activity and is suitable for research into solid tumors and lymphomas.
  • Potently inhibits proliferation of human tumor cell lines.
  • Suppresses hydroxyurea-induced H2AX phosphorylation.
  • Exhibits high selectivity against mTOR, DNA-PK, ATM, and PI3K.
  • Shows potent antiproliferative activity against various cancer cell lines.
  • Demonstrates potent anti-tumor efficacy in monotherapy in xenograft models of ovarian and colorectal cancer.
  • Causes complete tumor remission in mantle cell lymphoma models.
  • Has synergistic antitumor activity when combined with Carboplatin in platinum-resistant human CRC PDX models.
  • Exhibits moderate oral bioavailability and short elimination half-lives.