Learn More
Medchemexpress LLC Elimusertib-d3 | 00-00-0 | 98.9% | 378.45 | C20H18D3N7O | 1 MG
SDP
Supplier:  Medchemexpress LLC HY101566S1MG
Encompass_Preferred
Elimusertib-d3 is the deuterium-labeled analog of elimusertib, a potent and selective ATR kinase inhibitor. It is supplied as a high-purity analytical standard for research use, suitable for bioanalytical quantitation, metabolic profiling, and mechanistic investigations in oncology research.
  • Deuterium-labeled analog for use as an internal standard in mass spectrometry.
  • High purity (98.9%).
  • Suitable for bioanalytical quantitation and pharmacokinetic studies.
  • Useful for metabolic and mechanistic investigations.
  • Supplied in a small, defined quantity for analytical applications (1 mg).
  • Molecular formula C20H18D3N7O; molecular weight 378.45.
Catalog No. 50-002-49113