Learn More
Medchemexpress LLC Elimusertib-d3 (BAY 1895344-d3) | 00-00-0 | 98.9% | 378.45 g/mol | C20H18D3N7O | 10 MG
SDP
Supplier:  Medchemexpress LLC HY101566S10MG
Encompass_Preferred
Elimusertib-d3 is the deuterium-labeled analogue of a potent, orally active, and selective ATR inhibitor that demonstrates anti-tumor activity. It is supplied for laboratory research use only.
  • Deuterium-labeled analogue for tracer and mass-shift studies.
  • High purity suitable for analytical and preclinical research.
  • Solid form with light yellow to yellow appearance.
  • Powder storage stability: -20°C for long-term storage.
  • DMSO-soluble; ultrasonic assistance and freshly opened DMSO recommended for best solubility.
Catalog No. 50-002-47148