Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Elimusertib-d3 (BAY 1895344-d3) | 98.9% | 378.45 g/mol | C20H18D3N7O | 5 MG
SDP

Catalog No. 5000247905
Encompass_Preferred

Elimusertib-d3 is the deuterium-labeled analog of elimusertib, a potent, orally active, selective ATR inhibitor with antitumor activity. The deuterated compound is provided as a solid for research use, and is designed for use as a tracer or internal standard in quantitative analytical methods.

  • Deuterium-labeled internal standard for NMR, GC-MS, and LC-MS.
  • High purity: 98.9%.
  • Molecular formula C20H18D3N7O; molecular weight 378.45 g/mol.
  • Supplied as a solid powder in milligram-scale sizes.
  • Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).

Catalog No. 50-002-47905 Supplier Medchemexpress LLC Supplier No. HY101566S5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Elimusertib-d3 is the deuterium-labeled analog of elimusertib, a potent, orally active, selective ATR inhibitor with antitumor activity. The deuterated compound is provided as a solid for research use, and is designed for use as a tracer or internal standard in quantitative analytical methods.

  • Deuterium-labeled internal standard for NMR, GC-MS, and LC-MS.
  • High purity: 98.9%.
  • Molecular formula C20H18D3N7O; molecular weight 378.45 g/mol.
  • Supplied as a solid powder in milligram-scale sizes.
  • Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.