Learn More
Medchemexpress LLC Elimusertib hydrochloride | 00-00-0 | MFCD31561442 | 99.9% | 411.89 | C20H22ClN7O | 2 MG
SDP
Catalog No. 5000246047
Encompass_Preferred
Elimusertib hydrochloride is the hydrochloride salt of elimusertib, a potent, orally active ATR kinase inhibitor used for in vitro and preclinical research into DNA damage response and oncology. It is supplied as a research-grade solid or as ready-to-use DMSO solutions and is intended for laboratory research use only.
  • Potent ATR kinase inhibition at low-nanomolar concentrations.
  • High purity appropriate for research applications.
  • Available as solid and as 10 mM DMSO solutions for convenience.
  • Multiple pack sizes to support small-scale screening or larger studies.
  • Stable when stored sealed at 4°C; solvent stocks stable at -80°C or -20°C.
  • Yellow to orange solid allowing visual inspection on receipt.
  • Datasheet provided with handling and storage guidance.
Catalog No. 50-002-46047 Supplier Medchemexpress LLC Supplier No. HY101566A2MG
Elimusertib hydrochloride is the hydrochloride salt of elimusertib, a potent, orally active ATR kinase inhibitor used for in vitro and preclinical research into DNA damage response and oncology. It is supplied as a research-grade solid or as ready-to-use DMSO solutions and is intended for laboratory research use only.
  • Potent ATR kinase inhibition at low-nanomolar concentrations.
  • High purity appropriate for research applications.
  • Available as solid and as 10 mM DMSO solutions for convenience.
  • Multiple pack sizes to support small-scale screening or larger studies.
  • Stable when stored sealed at 4°C; solvent stocks stable at -80°C or -20°C.
  • Yellow to orange solid allowing visual inspection on receipt.
  • Datasheet provided with handling and storage guidance.