Learn More
Medchemexpress LLC Elimusertib hydrochloride | 00-00-0 | MFCD31561442 | 99.9% | 411.89 | C20H22ClN7O | 25 MG
SDP
Catalog No. 5000246046
Encompass_Preferred
Elimusertib hydrochloride is the hydrochloride salt of elimusertib, a potent and selective ATR kinase inhibitor used in preclinical oncology research. It exhibits low-nanomolar activity against ATR and is used to probe DNA damage response pathways, cell cycle effects, and anti-tumor activity in models of solid tumors and lymphomas. Supplied as a high-purity solid for laboratory experiments.
  • High purity (99.9%).
  • Potent, selective ATR inhibitor with low-nanomolar potency (IC50 ~7 nM).
  • Useful for DNA damage response and oncology research applications.
  • Supplied as a solid powder suitable for laboratory use.
  • Available in small pack sizes for preclinical studies, including 25 mg.
Catalog No. 50-002-46046 Supplier Medchemexpress LLC Supplier No. HY101566A25MG
Elimusertib hydrochloride is the hydrochloride salt of elimusertib, a potent and selective ATR kinase inhibitor used in preclinical oncology research. It exhibits low-nanomolar activity against ATR and is used to probe DNA damage response pathways, cell cycle effects, and anti-tumor activity in models of solid tumors and lymphomas. Supplied as a high-purity solid for laboratory experiments.
  • High purity (99.9%).
  • Potent, selective ATR inhibitor with low-nanomolar potency (IC50 ~7 nM).
  • Useful for DNA damage response and oncology research applications.
  • Supplied as a solid powder suitable for laboratory use.
  • Available in small pack sizes for preclinical studies, including 25 mg.