Learn More
Medchemexpress LLC Elimusertib hydrochloride | 00-00-0 | MFCD31561442 | 99.9% | 411.89 g/mol | C20H22ClN7O | 1 ML
SDP
Catalog No. 5000247919
Encompass_Preferred
Elimusertib hydrochloride is a selective ATR kinase inhibitor supplied for research use. It is available as a ready-to-use 10 mM solution in DMSO and as solid aliquots, commonly used in preclinical studies of DNA damage response and antitumor activity. Molecular formula: C20H22ClN7O; molecular weight: 411.89 g/mol.
  • Selective ATR kinase inhibitor for DNA damage response research.
  • Provided as a 10 mM solution in DMSO for ready-to-use assays.
  • Also available in solid form across multiple milligram pack sizes for assay development.
  • High purity (≈99.9%) suitable for biochemical and cellular assays.
  • Stable when stored sealed, protected from moisture, at recommended temperatures.
  • Manufacturer-provided data sheet and SDS available for handling and safety information.
Catalog No. 50-002-47919 Supplier Medchemexpress LLC Supplier No. HY101566A10MM 1ML
Elimusertib hydrochloride is a selective ATR kinase inhibitor supplied for research use. It is available as a ready-to-use 10 mM solution in DMSO and as solid aliquots, commonly used in preclinical studies of DNA damage response and antitumor activity. Molecular formula: C20H22ClN7O; molecular weight: 411.89 g/mol.
  • Selective ATR kinase inhibitor for DNA damage response research.
  • Provided as a 10 mM solution in DMSO for ready-to-use assays.
  • Also available in solid form across multiple milligram pack sizes for assay development.
  • High purity (≈99.9%) suitable for biochemical and cellular assays.
  • Stable when stored sealed, protected from moisture, at recommended temperatures.
  • Manufacturer-provided data sheet and SDS available for handling and safety information.