Learn More
Selleck Chemical LLC Elimusertib5mg
SDP
Supplier:  Selleck Chemical LLC S98645MG
Encompass_Preferred
Elimusertib (BAY-1895344) is a very potent and highly selectiveATR (ataxia telangiectasia and Rad3-related) inhibitor with IC50 of 7 nM. Elimusertib potently inhibits proliferation of a broad spectrum of human tumor cell lines with median IC50 of 78 nM. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-217-3513