Learn More
Pipette Supplies, Inc. ELINE BATTERY W/OUT COVER
SDP
Catalog No. 501317649
Encompass
Replacement battery pack for the Sartorius eLINE pipettes, without the plastic battery cover (requires reuse of your existing battery cover). Simply remove old batteries from your existing battery cover, and insert new battery pack.Rechargeable NiMH battery pack with protection circuit – 4.8 V, 500 mAH. Battery cells are 7/8in. in length. Battery must be charged prior to use.
Catalog No. 50-131-7649 Supplier Pipette Supplies, Inc. Supplier No. PS 731009
Replacement battery pack for the Sartorius eLINE pipettes, without the plastic battery cover (requires reuse of your existing battery cover). Simply remove old batteries from your existing battery cover, and insert new battery pack.Rechargeable NiMH battery pack with protection circuit – 4.8 V, 500 mAH. Battery cells are 7/8in. in length. Battery must be charged prior to use.