Learn More
Medchemexpress LLC HY-11021 5mg Medchemexpress, Elinogrel CAS:936500-94-6 Purity:>98%
SDP
Catalog No. 501968771
Encompass_Preferred
Medchemexpress, HY-11021 5mg Elinogrel CAS:936500-94-6 Elinogrel (PRT060128) is a potent, direct acting, competitive, and reversible platelet P2Y12 antagonist (IC50=20 nM). It is orally and intravenously available and has potent antiplatelet effects. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8771 Supplier Medchemexpress LLC Supplier No. HY110215MG
Medchemexpress, HY-11021 5mg Elinogrel CAS:936500-94-6 Elinogrel (PRT060128) is a potent, direct acting, competitive, and reversible platelet P2Y12 antagonist (IC50=20 nM). It is orally and intravenously available and has potent antiplatelet effects. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.