Learn More
Medchemexpress LLC Elinogrel | 936500-94-6 | 98.3% | 523.95 | 50 MG
SDP
Catalog No. 5000343698
Encompass_Preferred
Elinogrel (PRT060128) is a potent, direct acting, competitive, and reversible platelet P2Y12 antagonist. It is orally and intravenously available and has potent antiplatelet effects.
  • Potent, direct acting, competitive, and reversible platelet P2Y12 antagonist (IC50=20 nM)
  • Orally and intravenously available
  • Potent antiplatelet effects
Catalog No. 50-003-43698 Supplier Medchemexpress LLC Supplier No. HY1102110MG
Elinogrel (PRT060128) is a potent, direct acting, competitive, and reversible platelet P2Y12 antagonist. It is orally and intravenously available and has potent antiplatelet effects.
  • Potent, direct acting, competitive, and reversible platelet P2Y12 antagonist (IC50=20 nM)
  • Orally and intravenously available
  • Potent antiplatelet effects