Learn More
Selleck Chemical LLC Eliprodil
SDP
Catalog No. 5000000932
Encompass_Preferred
Eliprodil (SL-820715) is a non-competitive NR2B-selective NMDA antagonist with IC50 value of 1 M and is less potent for NR2A- and NR2C-containing receptors with IC50 100 M
Catalog No. 50-000-00932 Supplier Selleck Chemical LLC Supplier No. S67315MG
Eliprodil (SL-820715) is a non-competitive NR2B-selective NMDA antagonist with IC50 value of 1 M and is less potent for NR2A- and NR2C-containing receptors with IC50 100 M