Learn More
GVS FILTER TECHNOLOGY INC Elipse OV/P100 pair of filters for SPR466/467
Catalog No. NC1600849
Encompass_Preferred
This item has a minimum qty of 8 per supplier requirements.
Organic vapor cartridge has activated carbon with a pore structure for maximum absorption efficiency and a selected pore size for an optimised breathing resistance. Comes with two activated carbon filtering elements for the protection against a range of gases, vapors, dust, mist and fumes.
Catalog No. NC1600849 Supplier GVS FILTER TECHNOLOGY INC Supplier No. SPR468
This item has a minimum qty of 8 per supplier requirements.
Organic vapor cartridge has activated carbon with a pore structure for maximum absorption efficiency and a selected pore size for an optimised breathing resistance. Comes with two activated carbon filtering elements for the protection against a range of gases, vapors, dust, mist and fumes.