Learn More
GVS FILTER TECHNOLOGY INC Elipse P100 nuisance odour ready to use Mask with replaceable filters (S/M size)
Catalog No. NC1600838
Encompass_Preferred
This item has a minimum qty of 3 per supplier requirements.
GVS designed the Elipse respirators to fit the contours of the individuals face & the result is a form fitting respirator users will find very comfortable. Elipse masks also have been designed to completely seal around the nose area to prevent exhaled air from fogging the wearers safety glasses. low profile filter design offers a unique solution to customers by providing the user w/ a lightweight & comfortable respirator. Elipse provides wearers w/ a wide field of view & filters that don't interfere w/ other PPE such as sanding shields, safety glasses or welding hoods. The innovative close pleated composite P-100 filters provide users w/ the maximum particulate protection in a small & lightweight filter. Elipse respirators are re-usable & the intuitive design allows the filters to easily be interchanged. A low breathing resistance , 4 point harness for secure fit & soft thermoplastic elastomer mask material.
Catalog No. NC1600838 Supplier GVS FILTER TECHNOLOGY INC Supplier No. SPR449
This item has a minimum qty of 3 per supplier requirements.
GVS designed the Elipse respirators to fit the contours of the individuals face & the result is a form fitting respirator users will find very comfortable. Elipse masks also have been designed to completely seal around the nose area to prevent exhaled air from fogging the wearers safety glasses. low profile filter design offers a unique solution to customers by providing the user w/ a lightweight & comfortable respirator. Elipse provides wearers w/ a wide field of view & filters that don't interfere w/ other PPE such as sanding shields, safety glasses or welding hoods. The innovative close pleated composite P-100 filters provide users w/ the maximum particulate protection in a small & lightweight filter. Elipse respirators are re-usable & the intuitive design allows the filters to easily be interchanged. A low breathing resistance , 4 point harness for secure fit & soft thermoplastic elastomer mask material.