Learn More
Cayman Chemical Ellipticine (hydrochloride), Purity: ≥98%, CAS: 5081-48-1, 10 mg
SDP
Catalog No. 502967923
Encompass_Preferred
A DNA-intercalating antitumor agent that forms covalent adducts in DNA after being enzymatically activated with CYP3A4, CYP1A1, or CYP1A2 or by peroxidases in target tissues; inhibits the proliferation of human breast adenocarcinoma, leukemia, neuroblastoma, and glioblastoma cells (IC50s = 0.27-4.7 µM)
Catalog No. 50-296-7923 Supplier Cayman Chemical Supplier No. 1874210 MG
A DNA-intercalating antitumor agent that forms covalent adducts in DNA after being enzymatically activated with CYP3A4, CYP1A1, or CYP1A2 or by peroxidases in target tissues; inhibits the proliferation of human breast adenocarcinoma, leukemia, neuroblastoma, and glioblastoma cells (IC50s = 0.27-4.7 µM)