Learn More
Cayman Chemical Ellipticine (hydrochloride), Purity: ≥98%, CAS: 5081-48-1, 50 mg
SDP
Catalog No. 502966180
Encompass_Preferred
A DNA-intercalating antitumor agent that forms covalent adducts in DNA after being enzymatically activated with CYP3A4, CYP1A1, or CYP1A2 or by peroxidases in target tissues; inhibits the proliferation of human breast adenocarcinoma, leukemia, neuroblastoma, and glioblastoma cells (IC50s = 0.27-4.7 µM)
Catalog No. 50-296-6180 Supplier Cayman Chemical Supplier No. 1874250 MG
A DNA-intercalating antitumor agent that forms covalent adducts in DNA after being enzymatically activated with CYP3A4, CYP1A1, or CYP1A2 or by peroxidases in target tissues; inhibits the proliferation of human breast adenocarcinoma, leukemia, neuroblastoma, and glioblastoma cells (IC50s = 0.27-4.7 µM)