Learn More
BioVendor, LLC ELISA ADIPOCYTE FABP HUMAN (96 wells)
SDP
Supplier:  BioVendor, LLC RD191036200R
Encompass
TypeSandwich ELISA, Biotin-labelled antibodyApplicationsSerum, Plasma-EDTA, Plasma-Heparin, Plasma-CitrateSample Requirements20 µl/wellShippingAt ambient temperature. Upon receipt, store the product at the temperature recommended below.Storage/ExpirationStore the kit at 2–8°C. Under these conditions, the kit is stable until the expiration date (see label on the box). Calibration Range0.5–25 ng/mlLimit of Detection0.05 ng/ml
Catalog No. 50-894-197