Learn More
BioVendor, LLC Elisa Adiponectin Mouse BioVendor LLC
SDP
Supplier:  BioVendor, LLC RD293023100R
Encompass
TypeSandwich ELISA, Biotin-labelled antibodyApplicationsSerumSample Requirements10 ¿l/wellShippingAt ambient temperature. Upon receipt, store the product at the temperature recommended below.Storage/ExpirationStore the kit at 2¿8¿C. Under these conditions, the kit is stable until the expiration date (see label on the box). Calibration Range0.25¿8 ng/mlLimit of Detection0.079 ng/ml
Catalog No. 50-894-201